RIE156324799 Rieke Metals, Inc.

3-cyano-3'-pyrrolidinomethyl benzophenone

  • MDL number MFCD03842015
  • Cas Number : 898794-12-2
  • Molecular Weight : 290.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6125-0300-09

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0300-09
Availability details for
Quantity
  • Availability
  • Add to Cart