RIE156324800 Rieke Metals, Inc.

4'-cyano-3-pyrrolidinomethyl benzophenone

  • MDL number MFCD03842016
  • Cas Number : 898794-15-5
  • Molecular Weight : 290.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6125-0300-10

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0300-10
Availability details for
Quantity
  • Availability
  • Add to Cart