RIE156324807 Rieke Metals, Inc.

4'-bromo-3-pyrrolidinomethyl benzophenone

  • MDL number MFCD03842023
  • Cas Number : 898770-19-9
  • Molecular Weight : 344.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6125-0300-19

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0300-19
Availability details for
Quantity
  • Availability
  • Add to Cart