RIE156324812 Rieke Metals, Inc.

2,3-dimethyl-3'-pyrrolidinomethyl benzophenone

  • MDL number MFCD03842028
  • Cas Number : 898770-34-8
  • Molecular Weight : 293.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6125-0300-26

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0300-26
Availability details for
Quantity
  • Availability
  • Add to Cart