RIE156324816 Rieke Metals, Inc.

3,4-dimethyl-3'-pyrrolidinomethyl benzophenone

  • MDL number MFCD03842032
  • Cas Number : 898770-46-2
  • Molecular Weight : 293.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6125-0300-30

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0300-30
Availability details for
Quantity
  • Availability
  • Add to Cart