RIE156324818 Rieke Metals, Inc.

4-bromo-3-fluoro-3'-pyrrolidinomethyl benzophenone

  • MDL number MFCD03842034
  • Cas Number : 898770-52-0
  • Molecular Weight : 362.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6125-0300-32

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0300-32
Availability details for
Quantity
  • Availability
  • Add to Cart