RIE156324821 Rieke Metals, Inc.

2-chloro-3'-pyrrolidinomethyl benzophenone

  • MDL number MFCD03842037
  • Cas Number : 898770-58-6
  • Molecular Weight : 299.80

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6125-0300-36

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0300-36
Availability details for
Quantity
  • Availability
  • Add to Cart