RIE156324822 Rieke Metals, Inc.

2-fluoro-3'-pyrrolidinomethyl benzophenone

  • MDL number MFCD03842038
  • Cas Number : 898770-60-0
  • Molecular Weight : 283.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2-fluorophenyl)[3-(1-pyrrolidinylmethyl)phenyl]methanone
Inchi Code 1S/C18H18FNO/c19-17-9-2-1-8-16(17)18(21)15-7-5-6-14(12-15)13-20-10-3-4-11-20/h1-2,5-9,12H,3-4,10-11,13H2
Inchi Key ICWBFZHQCQGCFU-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6125-0300-37

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0300-37
Availability details for
Quantity
  • Availability
  • Add to Cart