RIE156324823 Rieke Metals, Inc.

3'-pyrrolidinomethyl-2-trifluoromethylbenzophenone

  • MDL number MFCD03842039
  • Cas Number : 898770-62-2
  • Molecular Weight : 333.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name [3-(1-pyrrolidinylmethyl)phenyl][2-(trifluoromethyl)phenyl]methanone
Inchi Code 1S/C19H18F3NO/c20-19(21,22)17-9-2-1-8-16(17)18(24)15-7-5-6-14(12-15)13-23-10-3-4-11-23/h1-2,5-9,12H,3-4,10-11,13H2
Inchi Key AQMWXLSDLIBTSL-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6125-0300-43

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0300-43
Availability details for
Quantity
  • Availability
  • Add to Cart