RIE156324832 Rieke Metals, Inc.

2,5-dichloro-3'-pyrrolidinomethyl benzophenone

  • MDL number MFCD03842048
  • Cas Number : 898770-80-4
  • Molecular Weight : 334.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6125-0300-55

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0300-55
Availability details for
Quantity
  • Availability
  • Add to Cart