RIE156324837 Rieke Metals, Inc.

3,5-difluoro-3'-pyrrolidinomethyl benzophenone

  • MDL number MFCD03842053
  • Cas Number : 898770-90-6
  • Molecular Weight : 301.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6125-0300-60

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0300-60
Availability details for
Quantity
  • Availability
  • Add to Cart