RIE156324838 Rieke Metals, Inc.

3'-pyrrolidinomethyl-3,4,5-trifluorobenzophenone

  • MDL number MFCD03842054
  • Cas Number : 898770-92-8
  • Molecular Weight : 319.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6125-0300-61

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0300-61
Availability details for
Quantity
  • Availability
  • Add to Cart