RIE156324840 Rieke Metals, Inc.

cyclobutyl 3-(pyrrolidinomethyl)phenyl ketone

  • MDL number MFCD03842056
  • Cas Number : 898770-96-2
  • Molecular Weight : 243.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name cyclobutyl[3-(1-pyrrolidinylmethyl)phenyl]methanone
Inchi Code 1S/C16H21NO/c18-16(14-6-4-7-14)15-8-3-5-13(11-15)12-17-9-1-2-10-17/h3,5,8,11,14H,1-2,4,6-7,9-10,12H2
Inchi Key FRULAKPZZZCKKX-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6125-0302-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0302-00
Availability details for
Quantity
  • Availability
  • Add to Cart