RIE156324843 Rieke Metals, Inc.

ethyl 4-oxo-4-[3-(pyrrolidinomethyl)phenyl]butyrate

  • MDL number MFCD03842059
  • Cas Number : 898771-02-3
  • Molecular Weight : 289.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 4-oxo-4-[3-(1-pyrrolidinylmethyl)phenyl]butanoate
Inchi Code 1S/C17H23NO3/c1-2-21-17(20)9-8-16(19)15-7-5-6-14(12-15)13-18-10-3-4-11-18/h5-7,12H,2-4,8-11,13H2,1H3
Inchi Key FXQNDXIGUZVPLC-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6125-0305-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0305-00
Availability details for
Quantity
  • Availability
  • Add to Cart