RIE156324846 Rieke Metals, Inc.

ethyl 7-oxo-7-[3-(pyrrolidinomethyl)phenyl]heptanoate

  • MDL number MFCD03842062
  • Cas Number : 898771-10-3
  • Molecular Weight : 331.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6125-0308-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0308-00
Availability details for
Quantity
  • Availability
  • Add to Cart