RIE156324851 Rieke Metals, Inc.

3-azetidinomethyl-4'-methylbenzophenone

  • MDL number MFCD03842067
  • Cas Number : 898771-25-0
  • Molecular Weight : 265.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6125-0400-04

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0400-04
Availability details for
Quantity
  • Availability
  • Add to Cart