RIE156324855 Rieke Metals, Inc.

3'-azetidinomethyl-2-cyanobenzophenone

  • MDL number MFCD03842071
  • Cas Number : 898771-36-3
  • Molecular Weight : 276.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-[3-(1-azetidinylmethyl)benzoyl]benzonitrile
Inchi Code 1S/C18H16N2O/c19-12-16-6-1-2-8-17(16)18(21)15-7-3-5-14(11-15)13-20-9-4-10-20/h1-3,5-8,11H,4,9-10,13H2
Inchi Key VBECEPWTMYZXDD-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6125-0400-08

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0400-08
Availability details for
Quantity
  • Availability
  • Add to Cart