RIE156324857 Rieke Metals, Inc.

3-azetidinomethyl-4'-cyanobenzophenone

  • MDL number MFCD03842073
  • Cas Number : 898771-42-1
  • Molecular Weight : 276.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6125-0400-10

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0400-10
Availability details for
Quantity
  • Availability
  • Add to Cart