RIE156324858 Rieke Metals, Inc.

3'-azetidinomethyl-2-carboethoxybenzophenone

  • MDL number MFCD03842074
  • Cas Number : 898771-45-4
  • Molecular Weight : 323.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 2-[3-(1-azetidinylmethyl)benzoyl]benzoate
Inchi Code 1S/C20H21NO3/c1-2-24-20(23)18-10-4-3-9-17(18)19(22)16-8-5-7-15(13-16)14-21-11-6-12-21/h3-5,7-10,13H,2,6,11-12,14H2,1H3
Inchi Key NOMYNSBGVWAJMS-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6125-0400-11

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0400-11
Availability details for
Quantity
  • Availability
  • Add to Cart