RIE156324859 Rieke Metals, Inc.

3-azetidinomethyl-3'-carboethoxybenzophenone

  • MDL number MFCD03842075
  • Cas Number : 898771-48-7
  • Molecular Weight : 323.40

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6125-0400-12

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0400-12
Availability details for
Quantity
  • Availability
  • Add to Cart