RIE156324862 Rieke Metals, Inc.

3-azetidinomethyl-4'-thiomethylbenzophenone

  • MDL number MFCD03842078
  • Cas Number : 898771-55-6
  • Molecular Weight : 297.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name [3-(1-azetidinylmethyl)phenyl][4-(methylsulfanyl)phenyl]methanone
Inchi Code 1S/C18H19NOS/c1-21-17-8-6-15(7-9-17)18(20)16-5-2-4-14(12-16)13-19-10-3-11-19/h2,4-9,12H,3,10-11,13H2,1H3
Inchi Key QOHKFSYIDJUCLV-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6125-0400-15

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0400-15
Availability details for
Quantity
  • Availability
  • Add to Cart