RIE156324863 Rieke Metals, Inc.

3-azetidinomethyl-3'-bromobenzophenone

  • MDL number MFCD03842079
  • Cas Number : 898771-57-8
  • Molecular Weight : 330.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6125-0400-18

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0400-18
Availability details for
Quantity
  • Availability
  • Add to Cart