RIE156324864 Rieke Metals, Inc.

3-azetidinomethyl-4'-bromobenzophenone

  • MDL number MFCD03842080
  • Cas Number : 898771-59-0
  • Molecular Weight : 330.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6125-0400-19

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0400-19
Availability details for
Quantity
  • Availability
  • Add to Cart