RIE156324866 Rieke Metals, Inc.

3-azetidinomethyl-4'-chlorobenzophenone

  • MDL number MFCD03842082
  • Cas Number : 898771-63-6
  • Molecular Weight : 285.78

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6125-0400-21

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0400-21
Availability details for
Quantity
  • Availability
  • Add to Cart