RIE156324871 Rieke Metals, Inc.

3'-azetidinomethyl-2,5-dimethylbenzophenone

  • MDL number MFCD03842087
  • Cas Number : 898771-73-8
  • Molecular Weight : 279.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6125-0400-28

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0400-28
Availability details for
Quantity
  • Availability
  • Add to Cart