RIE156324873 Rieke Metals, Inc.

3'-azetidinomethyl-3,4-dimethylbenzophenone

  • MDL number MFCD03842089
  • Cas Number : 898771-77-2
  • Molecular Weight : 279.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name [3-(1-azetidinylmethyl)phenyl](3,4-dimethylphenyl)methanone
Inchi Code 1S/C19H21NO/c1-14-7-8-18(11-15(14)2)19(21)17-6-3-5-16(12-17)13-20-9-4-10-20/h3,5-8,11-12H,4,9-10,13H2,1-2H3
Inchi Key INEZATBOKSMSEI-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6125-0400-30

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0400-30
Availability details for
Quantity
  • Availability
  • Add to Cart