RIE156324875 Rieke Metals, Inc.

3'-azetidinomethyl-4-bromo-3-fluorobenzophenone

  • MDL number MFCD03842091
  • Cas Number : 898771-81-8
  • Molecular Weight : 348.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name [3-(1-azetidinylmethyl)phenyl](4-bromo-3-fluorophenyl)methanone
Inchi Code 1S/C17H15BrFNO/c18-15-6-5-14(10-16(15)19)17(21)13-4-1-3-12(9-13)11-20-7-2-8-20/h1,3-6,9-10H,2,7-8,11H2
Inchi Key INPQEDYXWPSYGQ-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6125-0400-32

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0400-32
Availability details for
Quantity
  • Availability
  • Add to Cart