RIE156324877 Rieke Metals, Inc.

3'-azetidinomethyl-3-chloro-4-fluorobenzophenone

  • MDL number MFCD03842093
  • Cas Number : 898771-85-2
  • Molecular Weight : 303.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6125-0400-34

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0400-34
Availability details for
Quantity
  • Availability
  • Add to Cart