RIE156324882 Rieke Metals, Inc.

3-azetidinomethyl-4'-trifluoromethylbenzophenone

  • MDL number MFCD03842098
  • Cas Number : 898771-95-4
  • Molecular Weight : 319.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6125-0400-45

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0400-45
Availability details for
Quantity
  • Availability
  • Add to Cart