RIE156324883 Rieke Metals, Inc.

3'-azetidinomethyl-4-bromo-2-fluorobenzophenone

  • MDL number MFCD03842099
  • Cas Number : 898771-97-6
  • Molecular Weight : 348.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6125-0400-49

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0400-49
Availability details for
Quantity
  • Availability
  • Add to Cart