RIE156324884 Rieke Metals, Inc.

3'-azetidinomethyl-2-chloro-4-fluorobenzophenone

  • MDL number MFCD03842100
  • Cas Number : 898771-99-8
  • Molecular Weight : 303.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6125-0400-50

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0400-50
Availability details for
Quantity
  • Availability
  • Add to Cart