RIE156324885 Rieke Metals, Inc.

3'-azetidinomethyl-3-chloro-5-fluorobenzophenone

  • MDL number MFCD03842101
  • Cas Number : 898772-01-5
  • Molecular Weight : 303.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6125-0400-51

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0400-51
Availability details for
Quantity
  • Availability
  • Add to Cart