RIE156324888 Rieke Metals, Inc.

3'-azetidinomethyl-2,4-dichlorobenzophenone

  • MDL number MFCD03842104
  • Cas Number : 898772-09-3
  • Molecular Weight : 320.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6125-0400-54

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0400-54
Availability details for
Quantity
  • Availability
  • Add to Cart