RIE156324889 Rieke Metals, Inc.

3'-azetidinomethyl-2,5-dichlorobenzophenone

  • MDL number MFCD03842105
  • Cas Number : 898772-12-8
  • Molecular Weight : 320.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6125-0400-55

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0400-55
Availability details for
Quantity
  • Availability
  • Add to Cart