RIE156324890 Rieke Metals, Inc.

3'-azetidinomethyl-3,4-dichlorobenzophenone

  • MDL number MFCD03842106
  • Cas Number : 898772-15-1
  • Molecular Weight : 320.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name [3-(1-azetidinylmethyl)phenyl](3,4-dichlorophenyl)methanone
Inchi Code 1S/C17H15Cl2NO/c18-15-6-5-14(10-16(15)19)17(21)13-4-1-3-12(9-13)11-20-7-2-8-20/h1,3-6,9-10H,2,7-8,11H2
Inchi Key MRYKAJVKASFLBB-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6125-0400-56

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0400-56
Availability details for
Quantity
  • Availability
  • Add to Cart