RIE156324895 Rieke Metals, Inc.

3'-azetidinomethyl-3,4,5-trifluorobenzophenone

  • MDL number MFCD03842111
  • Cas Number : 898772-30-0
  • Molecular Weight : 305.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name [3-(1-azetidinylmethyl)phenyl](3,4,5-trifluorophenyl)methanone
Inchi Code 1S/C17H14F3NO/c18-14-8-13(9-15(19)16(14)20)17(22)12-4-1-3-11(7-12)10-21-5-2-6-21/h1,3-4,7-9H,2,5-6,10H2
Inchi Key PFQRRJVVCYEYAM-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6125-0400-61

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0400-61
Availability details for
Quantity
  • Availability
  • Add to Cart