RIE156324896 Rieke Metals, Inc.

3-(azetidinomethyl)phenyl cyclopropyl ketone

  • MDL number MFCD03842112
  • Cas Number : 898772-33-3
  • Molecular Weight : 215.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6125-0401-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0401-00
Availability details for
Quantity
  • Availability
  • Add to Cart