RIE156324897 Rieke Metals, Inc.

3-(azetidinomethyl)phenyl cyclobutyl ketone

  • MDL number MFCD03842113
  • Cas Number : 898772-36-6
  • Molecular Weight : 229.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6125-0402-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0402-00
Availability details for
Quantity
  • Availability
  • Add to Cart