RIE156324898 Rieke Metals, Inc.

3-(azetidinomethyl)phenyl cyclopentyl ketone

  • MDL number MFCD03842114
  • Cas Number : 898772-39-9
  • Molecular Weight : 243.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6125-0403-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0403-00
Availability details for
Quantity
  • Availability
  • Add to Cart