RIE156324900 Rieke Metals, Inc.

ethyl 4-[3-(azetidinomethyl)phenyl]-4-oxobutyrate

  • MDL number MFCD03842116
  • Cas Number : 898772-45-7
  • Molecular Weight : 275.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6125-0405-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0405-00
Availability details for
Quantity
  • Availability
  • Add to Cart