RIE156324901 Rieke Metals, Inc.

ethyl 5-[3-(azetidinomethyl)phenyl]-5-oxovalerate

  • MDL number MFCD03842117
  • Cas Number : 898772-48-0
  • Molecular Weight : 289.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6125-0406-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0406-00
Availability details for
Quantity
  • Availability
  • Add to Cart