RIE156324905 Rieke Metals, Inc.

ethyl 8-[3-(indolylmethyl)phenyl]-8-oxooctanoate

  • MDL number MFCD03842176
  • Cas Number : 898761-23-4
  • Molecular Weight : 391.51

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 8-[3-(1H-indol-1-ylmethyl)phenyl]-8-oxooctanoate
Inchi Code 1S/C25H29NO3/c1-2-29-25(28)15-6-4-3-5-14-24(27)22-12-9-10-20(18-22)19-26-17-16-21-11-7-8-13-23(21)26/h7-13,16-18H,2-6,14-15,19H2,1H3
Inchi Key VLFXIXRCFFHZOI-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6125-0509-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0509-00
Availability details for
Quantity
  • Availability
  • Add to Cart