RIE156324907 Rieke Metals, Inc.

3'-[8-(1,4-dioxa-8-azaspiro[4.5]decyl)methyl]-2-methyl benzophenone

  • MDL number MFCD03842178
  • Cas Number : 898761-29-0
  • Molecular Weight : 351.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name [3-(1,4-dioxa-8-azaspiro[4.5]dec-8-ylmethyl)phenyl](2-methylphenyl)methanone
Inchi Code 1S/C22H25NO3/c1-17-5-2-3-8-20(17)21(24)19-7-4-6-18(15-19)16-23-11-9-22(10-12-23)25-13-14-26-22/h2-8,15H,9-14,16H2,1H3
Inchi Key ZQHGJHYXDBQQSO-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6125-0600-02

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0600-02
Availability details for
Quantity
  • Availability
  • Add to Cart