RIE156324910 Rieke Metals, Inc.

3'-[8-(1,4-dioxa-8-azaspiro[4.5]decyl)methyl]-2-methoxy benzophenone

  • MDL number MFCD03842181
  • Cas Number : 898761-38-1
  • Molecular Weight : 367.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6125-0600-05

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0600-05
Availability details for
Quantity
  • Availability
  • Add to Cart