RIE156324917 Rieke Metals, Inc.

3-carboethoxy-3'-[8-(1,4-dioxa-8-azaspiro[4.5]decyl)methyl]benzophenone

  • MDL number MFCD03842188
  • Cas Number : 898761-59-6
  • Molecular Weight : 409.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6125-0600-12

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0600-12
Availability details for
Quantity
  • Availability
  • Add to Cart