RIE156324920 Rieke Metals, Inc.

3-[8-(1,4-dioxa-8-azaspiro[4.5]decyl)methyl]-4'-thiomethyl benzophenone

  • MDL number MFCD03842191
  • Cas Number : 898761-67-6
  • Molecular Weight : 383.51

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name [3-(1,4-dioxa-8-azaspiro[4.5]dec-8-ylmethyl)phenyl][4-(methylsulfanyl)phenyl]methanone
Inchi Code 1S/C22H25NO3S/c1-27-20-7-5-18(6-8-20)21(24)19-4-2-3-17(15-19)16-23-11-9-22(10-12-23)25-13-14-26-22/h2-8,15H,9-14,16H2,1H3
Inchi Key QAECQOFXHCVWIS-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6125-0600-15

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0600-15
Availability details for
Quantity
  • Availability
  • Add to Cart