RIE156324924 Rieke Metals, Inc.

4'-chloro-3-[8-(1,4-dioxa-8-azaspiro[4.5]decyl)methyl]benozphenone

  • MDL number MFCD03842195
  • Cas Number : 898761-75-6
  • Molecular Weight : 371.86

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (4-chlorophenyl)[3-(1,4-dioxa-8-azaspiro[4.5]dec-8-ylmethyl)phenyl]methanone
Inchi Code 1S/C21H22ClNO3/c22-19-6-4-17(5-7-19)20(24)18-3-1-2-16(14-18)15-23-10-8-21(9-11-23)25-12-13-26-21/h1-7,14H,8-13,15H2
Inchi Key NISADTRNAZOJOO-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6125-0600-21

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0600-21
Availability details for
Quantity
  • Availability
  • Add to Cart