RIE156324932 Rieke Metals, Inc.

3,5-dimethyl-3'-[8-(1,4-dioxa-8-azaspiro[4.5]decyl)methyl]benzophenone

  • MDL number MFCD03842203
  • Cas Number : 898761-91-6
  • Molecular Weight : 365.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6125-0600-31

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0600-31
Availability details for
Quantity
  • Availability
  • Add to Cart