RIE156324933 Rieke Metals, Inc.

4'-bromo-3-[8-(1,4-dioxa-8-azaspiro[4.5]decyl)methyl]-3'-fluorobenzophenone

  • MDL number MFCD07699566
  • Cas Number : 898761-93-8
  • Molecular Weight : 434.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (4-bromo-3-fluorophenyl)[3-(1,4-dioxa-8-azaspiro[4.5]dec-8-ylmethyl)phenyl]methanone
Inchi Code 1S/C21H21BrFNO3/c22-18-5-4-17(13-19(18)23)20(25)16-3-1-2-15(12-16)14-24-8-6-21(7-9-24)26-10-11-27-21/h1-5,12-13H,6-11,14H2
Inchi Key KOPDESVFBZDTLD-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6125-0600-32

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0600-32
Availability details for
Quantity
  • Availability
  • Add to Cart