RIE156324938 Rieke Metals, Inc.

3'-[8-(1,4-dioxa-8-azaspiro[4.5]decyl)methyl]-2-trifluorobenzophenone

  • MDL number MFCD03842208
  • Cas Number : 898762-03-3
  • Molecular Weight : 405.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6125-0600-43

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0600-43
Availability details for
Quantity
  • Availability
  • Add to Cart